Assay Details
| Gene Symbol: BACE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206062.3 | NP_001192991.1 | 5 - 6 | 870 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BACE2| Gene Symbol: BACE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206062.3 | NP_001192991.1 | 5 - 6 | 870 | 62 | |
Set Membership: |
|