Assay Details
| Gene Symbol: UXS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206064.1 | NP_001192993.1 | 9 - 10 | 799 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UXS1| Gene Symbol: UXS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206064.1 | NP_001192993.1 | 9 - 10 | 799 | 54 | |
Set Membership: |
|