Assay Details
| Gene Symbol: DYNC1LI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206152.1 | NP_001193081.1 | 4 - 5 | 533 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DYNC1LI2| Gene Symbol: DYNC1LI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206152.1 | NP_001193081.1 | 4 - 5 | 533 | 66 | |
Set Membership: |
|