Assay Details
| Gene Symbol: EPB41L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206169.1 | NP_001193098.1 | 21 - 22 | 3291 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPB41L3| Gene Symbol: EPB41L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206169.1 | NP_001193098.1 | 21 - 22 | 3291 | 57 | |
Set Membership: |
|