Assay Details
| Gene Symbol: DLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206170.3 | NP_001193099.1 | 1 - 2 | 125 | 67 | |
| GenBank mRNA | BC103372.1 | - | 1 - 2 | 112 | 67 | |
More Information
Set Membership: |
|

DLD
Genomic Map