Assay Details
| Gene Symbol: DNAAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206188.1 | NP_001193117.1 | 3 - 4 | 471 | 128 | |
| GenBank mRNA | BC118485.1 | - | 3 - 4 | 450 | 128 | |
More Information
Set Membership: |
|

DNAAF4
Genomic Map