Assay Details
| Gene Symbol: TRPV6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206189.1 | NP_001193118.1 | 1 - 2 | 340 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRPV6| Gene Symbol: TRPV6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206189.1 | NP_001193118.1 | 1 - 2 | 340 | 60 | |
Set Membership: |
|