Assay Details
| Gene Symbol: HAUS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206190.2 | NP_001193119.2 | 6 - 7 | 603 | 64 | |
| GenBank mRNA | BC119887.1 | - | 5 - 6 | 559 | 64 | |
More Information
Set Membership: |
|

HAUS4
Genomic Map