Assay Details
| Gene Symbol: LARGE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206192.1 | NP_001193121.1 | 9 - 10 | 1345 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LARGE2| Gene Symbol: LARGE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206192.1 | NP_001193121.1 | 9 - 10 | 1345 | 104 | |
Set Membership: |
|