Assay Details
| Gene Symbol: KLHL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206196.2 | NP_001193125.1 | 5 - 6 | 1458 | 66 | |
| GenBank mRNA | BC149170.1 | - | 5 - 6 | 1459 | 66 | |
More Information
Set Membership: |
|

KLHL24
Genomic Map