Assay Details
| Gene Symbol: DYNC2I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206205.1 | NP_001193134.1 | 3 - 4 | 550 | 89 | |
| GenBank mRNA | BC116094.1 | - | 3 - 4 | 504 | 89 | |
More Information
Set Membership: |
|

DYNC2I2
Genomic Map