Assay Details
| Gene Symbol: EXOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206213.1 | NP_001193142.1 | 5 - 6 | 646 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EXOG| Gene Symbol: EXOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206213.1 | NP_001193142.1 | 5 - 6 | 646 | 70 | |
Set Membership: |
|