Assay Details
Gene Symbol: TRIM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206256.3 | NP_001193185.1 | 2 - 3 | 499 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
