Assay Details
| Gene Symbol: CLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206264.2 | NP_001193193.1 | 12 - 13 | 1596 | 64 | |
| GenBank mRNA | BC149486.1 | - | 11 - 12 | 2343 | 64 | |
More Information
Set Membership: |
|

CLK2
Genomic Map