Assay Details
| Gene Symbol: YTHDF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192513.3 | NP_001179442.1 | 4 - 5 | 1982 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YTHDF3| Gene Symbol: YTHDF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192513.3 | NP_001179442.1 | 4 - 5 | 1982 | 87 | |
Set Membership: |
|