Assay Details
| Gene Symbol: KRT36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192525.2 | NP_001179454.2 | 6 - 7 | 1246 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT36| Gene Symbol: KRT36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192525.2 | NP_001179454.2 | 6 - 7 | 1246 | 85 | |
Set Membership: |
|