Assay Details
| Gene Symbol: TRIM64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192539.2 | NP_001179468.2 | 2 - 3 | 507 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM64| Gene Symbol: TRIM64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192539.2 | NP_001179468.2 | 2 - 3 | 507 | 104 | |
Set Membership: |
|