Assay Details
| Gene Symbol: TOPORS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192578.2 | NP_001179507.2 | 2 - 3 | 244 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOPORS| Gene Symbol: TOPORS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192578.2 | NP_001179507.2 | 2 - 3 | 244 | 90 | |
Set Membership: |
|