Assay Details
| Gene Symbol: GRB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192586.1 | NP_001179515.1 | 14 - 15 | 1692 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRB10| Gene Symbol: GRB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192586.1 | NP_001179515.1 | 14 - 15 | 1692 | 63 | |
Set Membership: |
|