Assay Details
| Gene Symbol: ETV3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192587.1 | NP_001179516.1 | 3 - 4 | 496 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ETV3L| Gene Symbol: ETV3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192587.1 | NP_001179516.1 | 3 - 4 | 496 | 58 | |
Set Membership: |
|