Assay Details
| Gene Symbol: MRE11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192605.1 | NP_001179534.1 | 17 - 18 | 2132 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRE11| Gene Symbol: MRE11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192605.1 | NP_001179534.1 | 17 - 18 | 2132 | 71 | |
Set Membership: |
|