Assay Details
Gene Symbol: GPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192634.2 | NP_001179563.2 | 2 - 3 | 171 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
