Assay Details
| Gene Symbol: CSF2RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192664.2 | NP_001179593.2 | 9 - 10 | 1357 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSF2RB| Gene Symbol: CSF2RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192664.2 | NP_001179593.2 | 9 - 10 | 1357 | 72 | |
Set Membership: |
|