Assay Details
| Gene Symbol: SH3BP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206288.3 | NP_001193217.1 | 1 - 2 | 144 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SH3BP5| Gene Symbol: SH3BP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206288.3 | NP_001193217.1 | 1 - 2 | 144 | 63 | |
Set Membership: |
|