Assay Details
| Gene Symbol: ZNF608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206306.3 | NP_001193235.2 | 2 - 3 | 1170 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF608| Gene Symbol: ZNF608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206306.3 | NP_001193235.2 | 2 - 3 | 1170 | 62 | |
Set Membership: |
|