Assay Details
| Gene Symbol: TNFRSF12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206327.1 | NP_001193256.1 | 2 - 3 | 225 | 105 | |
| NM_001206328.1 | NP_001193257.1 | 2 - 3 | 262 | 105 | ||
| GenBank mRNA | BF603409.1 | - | 2 - 3 | 220 | 105 | |
| BI536082.1 | - | 2 - 3 | 262 | 105 | ||
| DY222489.1 | - | 2 - 3 | 225 | 105 | ||
| GU594280.1 | - | 2 - 3 | 202 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206328.1 transcripts.
Set Membership: |
|

TNFRSF12A
Genomic Map