Assay Details
| Gene Symbol: PDIK1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206373.1 | NP_001193302.1 | 1 - 2 | 242 | 61 | |
| GenBank mRNA | BC112655.1 | - | 1 - 2 | 243 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206373.1 transcripts.
Set Membership: |
|

PDIK1L
Genomic Map