Assay Details
| Gene Symbol: ADH1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206387.1 | NP_001193316.1 | 5 - 6 | 638 | 96 | |
| GenBank mRNA | BC111290.1 | - | 5 - 6 | 638 | 96 | |
More Information
Set Membership: |
|

ADH1C
Genomic Map