Assay Details
| Gene Symbol: ADH1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206387.1 | NP_001193316.1 | 8 - 9 | 1176 | 87 | |
| GenBank mRNA | EV732637.1 | - | 1 - 2 | 297 | 87 | |
More Information
Set Membership: |
|

ADH1C
Genomic Map