Assay Details
| Gene Symbol: C1H3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206451.2 | NP_001193380.1 | 2 - 3 | 436 | 92 | |
| GenBank mRNA | BC120104.1 | - | 2 - 3 | 437 | 92 | |
More Information
Set Membership: |
|

C1H3orf38
Genomic Map