Assay Details
Gene Symbol: C25H16orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206478.1 | NP_001193407.1 | 1 - 2 | 57 | 90 | |
GenBank mRNA | BC109564.1 | - | 1 - 2 | 58 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206478.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
