Assay Details
| Gene Symbol: CRNKL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206522.2 | NP_001193451.2 | 5 - 6 | 724 | 70 | |
| GenBank mRNA | DT817337.1 | - | 5 - 6 | 724 | 70 | |
More Information
Set Membership: |
|

CRNKL1
Genomic Map