Assay Details
| Gene Symbol: MECP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206552.1 | NP_001193481.1 | 2 - 3 | 216 | 74 | |
| GenBank mRNA | BC148975.1 | - | 2 - 3 | 216 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206552.1 transcripts.
Set Membership: |
|

MECP2
Genomic Map