Assay Details
| Gene Symbol: MECP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206552.1 | NP_001193481.1 | 3 - 4 | 565 | 87 | |
| GenBank mRNA | EE347844.1 | - | 2 - 3 | 373 | 87 | |
More Information
Set Membership: |
|

MECP2
Genomic Map