Assay Details
| Gene Symbol: MRGBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206561.1 | NP_001193490.1 | 2 - 3 | 335 | 69 | |
| GenBank mRNA | EE344733.1 | - | 2 - 3 | 275 | 69 | |
More Information
Set Membership: |
|

MRGBP
Genomic Map