Assay Details
| Gene Symbol: ALDH5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192735.1 | NP_001179664.1 | 6 - 7 | 982 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALDH5A1| Gene Symbol: ALDH5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192735.1 | NP_001179664.1 | 6 - 7 | 982 | 58 | |
Set Membership: |
|