Assay Details
| Gene Symbol: RAB11FIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192771.1 | NP_001179700.1 | 4 - 5 | 1605 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB11FIP2| Gene Symbol: RAB11FIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192771.1 | NP_001179700.1 | 4 - 5 | 1605 | 75 | |
Set Membership: |
|