Assay Details
Gene Symbol: ADAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001354175.1 | NP_001341104.1 | 6 - 7 | 734 | 66 | |
GenBank mRNA | DN640847.1 | - | 4 - 5 | 411 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
