Assay Details
| Gene Symbol: SALL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192836.3 | NP_001179765.3 | 3 - 4 | 2762 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SALL4| Gene Symbol: SALL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192836.3 | NP_001179765.3 | 3 - 4 | 2762 | 72 | |
Set Membership: |
|