Assay Details
| Gene Symbol: TRIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192840.2 | NP_001179769.2 | 10 - 11 | 1233 | 78 | |
| GenBank mRNA | BC114671.1 | - | 10 - 11 | 1292 | 78 | |
More Information
Set Membership: |
|

TRIT1
Genomic Map