Assay Details
| Gene Symbol: ANAPC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192845.1 | NP_001179774.1 | 3 - 4 | 503 | 75 | |
| GenBank mRNA | BC123612.1 | - | 3 - 4 | 478 | 75 | |
More Information
Set Membership: |
|

ANAPC7
Genomic Map