Assay Details
| Gene Symbol: GALNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192858.3 | NP_001179787.3 | 8 - 9 | 1625 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GALNT3| Gene Symbol: GALNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192858.3 | NP_001179787.3 | 8 - 9 | 1625 | 67 | |
Set Membership: |
|