Assay Details
| Gene Symbol: TRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163943.1 | NP_001157415.1 | 4 - 5 | 357 | 112 | |
| GenBank mRNA | BC103345.1 | - | 4 - 5 | 357 | 112 | |
More Information
Set Membership: |
|

TRAT1
Genomic Map