Assay Details
| Gene Symbol: CDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166492.3 | NP_001159964.3 | 13 - 14 | 2598 | 64 | |
| GenBank mRNA | X53615.1 | - | 2124 | 64 | ||
More Information
Set Membership: |
|

CDH2
Genomic Map