Assay Details
| Gene Symbol: ALDH3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166513.1 | NP_001159985.1 | 1 - 2 | 179 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALDH3A1| Gene Symbol: ALDH3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166513.1 | NP_001159985.1 | 1 - 2 | 179 | 101 | |
Set Membership: |
|