Assay Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206596.1 | NP_001193525.1 | 7 - 8 | 2246 | 66 | |
| GenBank mRNA | BC151750.1 | - | 7 - 8 | 2247 | 66 | |
| DV870488.1 | - | 1 - 2 | 32 | 66 | ||
More Information
Set Membership: |
|

PTPDC1
Genomic Map