Assay Details
| Gene Symbol: DVL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206601.1 | NP_001193530.1 | 1 - 2 | 168 | 83 | |
| GenBank mRNA | BC154382.1 | - | 1 - 2 | 127 | 83 | |
More Information
Set Membership: |
|

DVL1
Genomic Map