Assay Details
| Gene Symbol: SERPINB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206610.1 | NP_001193539.1 | 4 - 5 | 565 | 101 | |
| GenBank mRNA | BT025443.1 | - | 5 - 6 | 582 | 101 | |
More Information
Set Membership: |
|

SERPINB1
Genomic Map