Assay Details
| Gene Symbol: KLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206636.1 | NP_001193565.1 | 4 - 5 | 437 | 98 | |
| GenBank mRNA | AF322886.1 | - | 4 - 5 | 379 | 98 | |
More Information
Set Membership: |
|

KLRB1
Genomic Map