Assay Details
| Gene Symbol: SIRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206669.1 | NP_001193598.1 | 1 - 2 | 129 | 96 | |
| GenBank mRNA | BC149729.1 | - | 1 - 2 | 108 | 96 | |
| EV693057.1 | - | 1 - 2 | 134 | 96 | ||
More Information
Set Membership: |
|

SIRT3
Genomic Map